Assay Details
| Gene Symbol: Stard5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192011.1 | NP_001178940.1 | 3 - 4 | 330 | 78 | |
| GenBank mRNA | FQ230578.1 | - | 3 - 4 | 304 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stard5
Genomic Map