Assay Details
| Gene Symbol: Abhd17c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100736.1 | NP_001094206.1 | 1 - 2 | 573 | 139 | |
| GenBank mRNA | BC169098.1 | - | 1 - 2 | 573 | 7939241 | 139 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Abhd17c
Genomic Map