Assay Details
| Gene Symbol: Poteg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014238.1 | NP_001014260.1 | 5 - 6 | 903 | 250 | |
| XM_008771330.1 | XP_008769552.1 | 5 - 6 | 952 | 250 | ||
| GenBank mRNA | BC079270.1 | - | 5 - 6 | 903 | 7133714 | 250 |
More Information
Updated Mapping Information
Set Membership: |
|

Poteg
Genomic Map