Assay Details
| Gene Symbol: Fgf19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130753.1 | NP_570109.1 | 1 - 2 | 252 | 91 | |
| GenBank mRNA | AB078900.1 | - | 1 - 2 | 252 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgf19
Genomic Map