Assay Details
| Gene Symbol: Zdhhc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039100.1 | NP_001034189.1 | 1 - 2 | 300 | 120 | |
| GenBank mRNA | AY886540.1 | - | 1 - 2 | 300 | 120 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zdhhc24
Genomic Map