Assay Details
| Gene Symbol: Rassf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106317.2 | NP_001099787.1 | 4 - 5 | 1051 | 65 | |
| XM_006230537.2 | XP_006230599.1 | 5 - 6 | 1151 | 65 | ||
| XM_006230538.2 | XP_006230600.1 | 5 - 6 | 1444 | 65 | ||
| XM_006230539.2 | XP_006230601.1 | 4 - 5 | 1047 | 65 | ||
| GenBank mRNA | BC158743.1 | - | 4 - 5 | 1051 | 9100666 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Rassf7
Genomic Map