Assay Details
| Gene Symbol: Rassf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230538.2 | XP_006230600.1 | 1 - 2 | 394 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006230538.2 transcripts.
Set Membership: |
|

Rassf7
Genomic Map