Assay Details
| Gene Symbol: Hvcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249369.2 | XP_006249431.1 | 2 - 3 | 1525 | 70 | |
| XM_008769280.1 | XP_008767502.1 | 5 - 6 | 692 | 70 | ||
More Information
Set Membership: |
|

Hvcn1
Genomic Map