Assay Details
| Gene Symbol: Nyap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107132.1 | NP_001100602.1 | 4 - 5 | 2576 | 123 | |
| XM_006249149.2 | XP_006249211.1 | 3126 | 123 | |||
| Gene Symbol: LOC100910838 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006249089.2 | XP_006249151.1 | 3 - 4 | 3126 | 123 | |
| Gene Symbol: LOC103690980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008758797.1 | XP_008757019.1 | 1 - 2 | 286 | 123 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nyap1
Genomic Map