Assay Details
| Gene Symbol: Nsun5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191593.1 | NP_001178522.1 | 6 - 7 | 748 | 66 | |
| XM_008769129.1 | XP_008767351.1 | 6 - 7 | 800 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nsun5
Genomic Map