Assay Details
| Gene Symbol: Mboat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107317.2 | NP_001100787.2 | 2 | 95 | ||
| GenBank mRNA | EU518497.1 | - | 2 | 95 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mboat4
Genomic Map