Assay Details
| Gene Symbol: Got1l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014003.1 | NP_001014025.1 | 8 - 9 | 1058 | 107 | |
| GenBank mRNA | BC079187.1 | - | 8 - 9 | 1058 | 7131193 | 107 |
More Information
Updated Mapping Information
Set Membership: |
|

Got1l1
Genomic Map