Assay Details
| Gene Symbol: Tex15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106087.1 | NP_001099557.1 | 1 - 2 | 7053 | 86 | |
| XM_006253218.2 | XP_006253280.1 | 8 - 9 | 8100 | 86 | ||
| XM_006253221.2 | XP_006253283.1 | 5 - 6 | 7603 | 86 | ||
| XM_008771296.1 | XP_008769518.1 | 8 - 9 | 8072 | 86 | ||
| XM_008771297.1 | XP_008769519.1 | 8 - 9 | 8116 | 86 | ||
| XM_008771298.1 | XP_008769520.1 | 7 - 8 | 7940 | 86 | ||
| XM_008771299.1 | XP_008769521.1 | 4 - 5 | 7529 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tex15
Genomic Map