Assay Details
| Gene Symbol: Arglu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024972.1 | NP_001020143.1 | 2 - 3 | 772 | 77 | |
| GenBank mRNA | BC091344.1 | - | 2 - 3 | 772 | 7315932 | 77 |
| BC127459.1 | - | 2 - 3 | 729 | 7125115 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arglu1
Genomic Map