Assay Details
| Gene Symbol: C4bpb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016995.2 | NP_058691.2 | 3 - 4 | 684 | 105 | |
| XM_006249705.2 | XP_006249767.1 | 4 - 5 | 740 | 105 | ||
| XM_008769423.1 | XP_008767645.1 | 4 - 5 | 557 | 105 | ||
| GenBank mRNA | BC088152.1 | - | 3 - 4 | 684 | 7322205 | 105 |
| FQ209636.1 | - | 3 - 4 | 695 | 105 | ||
| FQ218474.1 | - | 3 - 4 | 665 | 105 | ||
| Z50052.1 | - | 3 - 4 | 663 | 105 | ||
More Information
Updated Mapping Information
Set Membership: |
|

C4bpb
Genomic Map