Assay Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109069.1 | NP_001102539.1 | 8 - 9 | 1524 | 76 | |
| XM_006249791.2 | XP_006249853.1 | 8 - 9 | 1519 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mfsd4
Genomic Map