Assay Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013126.1 | NP_001013144.1 | 2 - 3 | 208 | 60 | |
| GenBank mRNA | BC089945.1 | - | 2 - 3 | 208 | 7311492 | 60 |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r1
Genomic Map