Assay Details
| Gene Symbol: Cyb5r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013126.1 | NP_001013144.1 | 4 - 5 | 381 | 117 | |
| GenBank mRNA | BC089945.1 | - | 4 - 5 | 381 | 7311492 | 117 |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r1
Genomic Map