Assay Details
| Gene Symbol: Atp5g3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006234393.2 | XP_006234455.1 | 2 - 3 | 210 | 134 | |
| XM_006234394.2 | XP_006234456.1 | 2 - 3 | 190 | 134 | ||
| GenBank mRNA | FQ217141.1 | - | 2 - 3 | 148 | 134 | |
| FQ223732.1 | - | 2 - 3 | 158 | 134 | ||
| FQ224352.1 | - | 1 - 2 | 116 | 134 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006234393.2, XM_006234394.2 transcripts.
Set Membership: |
|

Atp5g3
Genomic Map