Assay Details
| Gene Symbol: Slc5a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108588.1 | NP_001102058.1 | 13 - 14 | 2021 | 60 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P112304 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P063601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591126 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816485 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404974 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529879 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147302 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6502 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P112304 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P063601 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P112304 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P063601 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P063601 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P112304 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P112304 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10838396 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838392 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838389 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838390 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838384 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838388 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838391 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838398 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838386 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838397 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838385 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838395 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838394 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838387 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10838383 | - | - |
| Rat Genome 230 2.0 Array | 1383832_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1391533_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1383832_at | - | - |
| Rat Genome 230 2.0 Array | 1391533_at | - | - |
| Affymetrix HT Rat Focus Array | 1391533_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1383832_at | - | - |
| Affymetrix HT Rat Focus Array | 1391533_at | - | - |
| Affymetrix HT Rat Focus Array | 1383832_PM_at | - | - |
| Rat Expression Set 230 | 1391533_at | - | - |
| Rat Expression Set 230 | 1383832_PM_at | - | - |
| Rat Expression Set 230 | 1391533_PM_at | - | - |
| Rat Expression Set 230 | 1383832_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10838383 | - | - |
| Rat Genome U34 Set | rc_AI058978_at | - | - |
| Rat Genome U34 Set | rc_AI073075_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21496812 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357362 | - | - |

Slc5a12
Genomic Map