Assay Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191853.1 | NP_001178782.1 | 3 - 4 | 324 | 119 | |
| XM_006234653.2 | XP_006234715.1 | 3 - 4 | 368 | 119 | ||
| XM_006234654.2 | XP_006234716.1 | 3 - 4 | 357 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc24
Genomic Map