Assay Details
| Gene Symbol: Rapsn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108584.1 | NP_001102054.1 | 5 - 6 | 970 | 84 | |
| XM_006234528.2 | XP_006234590.1 | 6 - 7 | 1520 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rapsn
Genomic Map