Assay Details
| Gene Symbol: Akr1e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008342.1 | NP_001008343.1 | 1 - 2 | 104 | 102 | |
| XM_008771865.1 | XP_008770087.1 | 1 - 2 | 124 | 102 | ||
| GenBank mRNA | BC086397.1 | - | 1 - 2 | 104 | 7302893 | 102 |
| FQ212583.1 | - | 1 - 2 | 117 | 102 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC086397.1,NM_001008342.1,XM_008771865.1 transcripts.
Set Membership: |
|

Akr1e2
Genomic Map