Assay Details
| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771881.1 | XP_008770103.1 | 1 - 2 | 581 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam107b| Gene Symbol: Fam107b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008771881.1 | XP_008770103.1 | 1 - 2 | 581 | 89 | |
Set Membership: |
|