Assay Details
| Gene Symbol: Il15ra | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728509.4 | XP_002728555.2 | 5 - 6 | 1204 | 57 | |
| XM_006254225.2 | XP_006254287.1 | 4 - 5 | 1001 | 57 | ||
More Information
Set Membership: |
|

Il15ra
Genomic Map