Assay Details
| Gene Symbol: Dhtkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025720.1 | NP_001020891.1 | 4 - 5 | 791 | 63 | |
| GenBank mRNA | BC099075.1 | - | 4 - 5 | 791 | 7441857 | 63 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P665075 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939020 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243680 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 532328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582137 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711982 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844674 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322613 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721756 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122650 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694840 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403621 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910941 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375910 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140715 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P665075 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P665075 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P665075 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P665075 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P475819 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17039 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 16203 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96329 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93737 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46469 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 700 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10796278 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796266 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796274 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796268 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796264 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796273 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796279 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796277 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796276 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796265 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796270 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796269 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796271 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796267 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796263 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796275 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796272 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10796262 | - | - |
| Rat Genome 230 2.0 Array | 1390172_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1390172_at | - | - |
| Affymetrix HT Rat Focus Array | 1390172_at | - | - |
| Affymetrix HT Rat Focus Array | 1390172_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1700006732.rn.1 | TC1700000984.rn.1 | RTA TC |
| Rat Expression Set 230 | 1390172_at | - | - |
| Rat Expression Set 230 | 1390172_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10796262 | - | - |
| Rat Genome U34 Set | rc_AI169947_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21111354 | - | - |
| ABI Rat Genome Survey Microarray | 21322967 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1348881 | - | - |

Dhtkd1
Genomic Map