Assay Details
| Gene Symbol: Atp5c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053825.2 | NP_446277.1 | 3 - 4 | 258 | 64 | |
| XM_006254229.2 | XP_006254291.1 | 3 - 4 | 312 | 64 | ||
| XM_006254230.2 | XP_006254292.1 | 3 - 4 | 313 | 64 | ||
| XM_006254231.2 | XP_006254293.1 | 3 - 4 | 224 | 64 | ||
| GenBank mRNA | BC059158.1 | - | 3 - 4 | 262 | 6920493 | 64 |
| FQ212577.1 | - | 3 - 4 | 246 | 64 | ||
| FQ212651.1 | - | 3 - 4 | 256 | 64 | ||
| FQ212813.1 | - | 3 - 4 | 256 | 64 | ||
| FQ213008.1 | - | 3 - 4 | 256 | 64 | ||
| FQ213785.1 | - | 3 - 4 | 258 | 64 | ||
| FQ213852.1 | - | 3 - 4 | 258 | 64 | ||
| FQ213883.1 | - | 3 - 4 | 252 | 64 | ||
| FQ214745.1 | - | 3 - 4 | 236 | 64 | ||
| FQ214863.1 | - | 3 - 4 | 312 | 64 | ||
| FQ215205.1 | - | 3 - 4 | 256 | 64 | ||
| FQ215286.1 | - | 3 - 4 | 231 | 64 | ||
| FQ215426.1 | - | 3 - 4 | 255 | 64 | ||
| FQ215847.1 | - | 3 - 4 | 256 | 64 | ||
| FQ216198.1 | - | 3 - 4 | 252 | 64 | ||
| FQ216285.1 | - | 3 - 4 | 246 | 64 | ||
| FQ216490.1 | - | 3 - 4 | 256 | 64 | ||
| FQ216539.1 | - | 3 - 4 | 240 | 64 | ||
| FQ216647.1 | - | 3 - 4 | 258 | 64 | ||
| FQ216736.1 | - | 3 - 4 | 256 | 64 | ||
| FQ218995.1 | - | 3 - 4 | 258 | 64 | ||
| FQ219484.1 | - | 3 - 4 | 256 | 64 | ||
| FQ219563.1 | - | 3 - 4 | 256 | 64 | ||
| FQ220076.1 | - | 3 - 4 | 256 | 64 | ||
| FQ220217.1 | - | 3 - 4 | 258 | 64 | ||
| FQ220355.1 | - | 3 - 4 | 256 | 64 | ||
| FQ224939.1 | - | 3 - 4 | 256 | 64 | ||
| FQ229987.1 | - | 3 - 4 | 279 | 64 | ||
| FQ234981.1 | - | 3 - 4 | 256 | 64 | ||
| L19927.1 | - | 1 - 2 | 136 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atp5c1
Genomic Map