Assay Details
| Gene Symbol: Atp5c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053825.2 | NP_446277.1 | 4 - 5 | 464 | 103 | |
| XM_006254229.2 | XP_006254291.1 | 4 - 5 | 518 | 103 | ||
| XM_006254230.2 | XP_006254292.1 | 4 - 5 | 519 | 103 | ||
| XM_006254231.2 | XP_006254293.1 | 4 - 5 | 430 | 103 | ||
| GenBank mRNA | BC059158.1 | - | 4 - 5 | 468 | 6920493 | 103 |
| FQ212577.1 | - | 4 - 5 | 452 | 103 | ||
| FQ212651.1 | - | 4 - 5 | 462 | 103 | ||
| FQ212813.1 | - | 4 - 5 | 462 | 103 | ||
| FQ213008.1 | - | 4 - 5 | 462 | 103 | ||
| FQ213785.1 | - | 4 - 5 | 464 | 103 | ||
| FQ213852.1 | - | 4 - 5 | 464 | 103 | ||
| FQ213883.1 | - | 4 - 5 | 458 | 103 | ||
| FQ214745.1 | - | 4 - 5 | 442 | 103 | ||
| FQ214863.1 | - | 4 - 5 | 518 | 103 | ||
| FQ214998.1 | - | 4 - 5 | 462 | 103 | ||
| FQ215205.1 | - | 4 - 5 | 462 | 103 | ||
| FQ215286.1 | - | 4 - 5 | 437 | 103 | ||
| FQ215351.1 | - | 4 - 5 | 462 | 103 | ||
| FQ215426.1 | - | 4 - 5 | 461 | 103 | ||
| FQ215847.1 | - | 4 - 5 | 462 | 103 | ||
| FQ216198.1 | - | 4 - 5 | 458 | 103 | ||
| FQ216285.1 | - | 4 - 5 | 452 | 103 | ||
| FQ216490.1 | - | 4 - 5 | 462 | 103 | ||
| FQ216539.1 | - | 4 - 5 | 446 | 103 | ||
| FQ216736.1 | - | 4 - 5 | 462 | 103 | ||
| FQ218995.1 | - | 4 - 5 | 464 | 103 | ||
| FQ219484.1 | - | 4 - 5 | 462 | 103 | ||
| FQ219563.1 | - | 4 - 5 | 462 | 103 | ||
| FQ219689.1 | - | 4 - 5 | 464 | 103 | ||
| FQ220076.1 | - | 4 - 5 | 462 | 103 | ||
| FQ220217.1 | - | 4 - 5 | 464 | 103 | ||
| FQ220355.1 | - | 4 - 5 | 462 | 103 | ||
| FQ224939.1 | - | 4 - 5 | 462 | 103 | ||
| FQ229987.1 | - | 4 - 5 | 485 | 103 | ||
| FQ234981.1 | - | 4 - 5 | 462 | 103 | ||
| L19927.1 | - | 2 - 3 | 342 | 103 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Atp5c1
Genomic Map