Assay Details
| Gene Symbol: Akr1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033697.1 | NP_001028869.1 | 7 - 8 | 861 | 136 | |
| GenBank mRNA | AB300410.1 | - | 7 - 8 | 852 | 136 | |
| BC088227.1 | - | 7 - 8 | 850 | 136 | ||
| BC104716.1 | - | 7 - 8 | 861 | 7373256 | 136 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akr1c1
Genomic Map