Assay Details
| Gene Symbol: Irf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047086.1 | NP_001040551.1 | 8 - 9 | 851 | 57 | |
| XM_006253139.2 | XP_006253201.1 | 8 - 9 | 1112 | 57 | ||
| XM_008771256.1 | XP_008769478.1 | 8 - 9 | 947 | 57 | ||
| GenBank mRNA | BC166593.1 | - | 8 - 9 | 892 | 7934031 | 57 |
| DQ674266.1 | - | 8 - 9 | 851 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulatory region DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
regulatory region DNA binding
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P073555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817342 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15398 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376945 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402791 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371018 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207460 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299003 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835821 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812306 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696802 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430708 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843881 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50732 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896833 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397678 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152728 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 143330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131384 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150096 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073555 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P073555 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P073555 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P466417 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17525 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85108 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44339 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8094 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10791643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791646 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791641 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791644 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10791640 | - | - |
| Rat Genome 230 2.0 Array | 1398561_at | - | - |
| Rat Genome 230 2.0 Array | 1398561_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382503_at | - | - |
| Rat Genome 230 2.0 Array | 1386518_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1382503_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1386518_at | - | - |
| Affymetrix HT Rat Focus Array | 1386518_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1386518_at | - | - |
| Affymetrix HT Rat Focus Array | 1382503_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1382503_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600012218.rn.1 | TC1600001640.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600012216.rn.1 | TC1600001640.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600012217.rn.1 | TC1600001640.rn.1 | RTA TC |
| Rat Expression Set 230 | 1386518_PM_at | - | - |
| Rat Expression Set 230 | 1382503_at | - | - |
| Rat Expression Set 230 | 1382503_PM_at | - | - |
| Rat Expression Set 230 | 1386518_at | - | - |
| Rat Expression Set 230 | 1398561_PM_at | - | - |
| Rat Expression Set 230 | 1398561_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791640 | - | - |
| Rat Genome U34 Set | AB003091_at | - | - |
| Rat Genome U34 Set | rc_AI102062_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20774341 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374809 | - | - |

Irf2
Genomic Map