Assay Details
| Gene Symbol: Slc25a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053515.1 | NP_445967.1 | 2 - 3 | 712 | 83 | |
| GenBank mRNA | BC060533.1 | - | 2 - 3 | 716 | 6922056 | 83 |
| D12770.1 | - | 2 - 3 | 679 | 83 | ||
| FQ213027.1 | - | 2 - 3 | 709 | 83 | ||
| FQ213086.1 | - | 2 - 3 | 709 | 83 | ||
| FQ213612.1 | - | 2 - 3 | 738 | 83 | ||
| FQ214384.1 | - | 2 - 3 | 708 | 83 | ||
| FQ220525.1 | - | 2 - 3 | 707 | 83 | ||
| FQ224814.1 | - | 2 - 3 | 709 | 83 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Slc25a4
Genomic Map