Assay Details
| Gene Symbol: Enpp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107311.1 | NP_001100781.1 | 6 - 7 | 1017 | 75 | |
| XM_006253156.2 | XP_006253218.1 | 6 - 7 | 1071 | 75 | ||
| XM_008771259.1 | XP_008769481.1 | 6 - 7 | 1071 | 75 | ||
| GenBank mRNA | BC158772.1 | - | 6 - 7 | 1033 | 7128690 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P058107 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647082 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717371 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251906 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723396 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745628 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279363 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469970 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 597500 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590187 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728325 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675676 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 183762 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37768 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505980 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465704 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710734 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875891 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947694 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 93090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 100565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1061 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 25231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27277 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169317 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058107 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P058107 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P058107 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15051 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59072 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 38068 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62719 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10791633 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791632 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791635 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791634 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791636 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791638 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10791631 | - | - |
| Rat Genome 230 2.0 Array | 1391864_at | - | - |
| Rat Genome 230 2.0 Array | 1391864_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391864_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1391864_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1600012198.rn.1 | TC1600001636.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1600012196.rn.1 | TC1600001636.rn.1 | RTA TC |
| Rat Expression Set 230 | 1391864_PM_at | - | - |
| Rat Expression Set 230 | 1391864_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10791631 | - | - |
| Rat Genome U34 Set | rc_AA891719_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21041330 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368726 | - | - |

Enpp6
Genomic Map