Assay Details
| Gene Symbol: N6amt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134990.1 | NP_001128462.1 | 1 - 2 | 167 | 76 | |
| XM_006252059.2 | XP_006252121.1 | 2 - 3 | 1020 | 76 | ||
| GenBank mRNA | BC168660.1 | - | 1 - 2 | 167 | 5621479 | 76 |
| FQ230419.1 | - | 2 - 3 | 217 | 76 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC168660.1,NM_001134990.1 transcripts.
Set Membership: |
|

N6amt2
Genomic Map