Assay Details
| Gene Symbol: Dhrs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001078405.6 | XP_001078405.4 | 6 - 7 | 622 | 60 | |
| XM_008770715.1 | XP_008768937.1 | 6 - 7 | 638 | 60 | ||
More Information
Set Membership: |
|

Dhrs2
Genomic Map