Assay Details
| Gene Symbol: Fgf9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012952.1 | NP_037084.1 | 2 - 3 | 532 | 111 | |
| GenBank mRNA | BC078699.1 | - | 1135 | 111 | ||
| D14839.1 | - | 2 - 3 | 551 | 111 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
osteoblast differentiation
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
protein import into nucleus
cell-cell signaling
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
male gonad development
positive regulation of gene expression
substantia nigra development
negative regulation of Wnt signaling pathway
male sex determination
lung development
embryonic limb morphogenesis
positive regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of activin receptor signaling pathway
inner ear morphogenesis
positive regulation of MAPK cascade
positive regulation of smoothened signaling pathway
regulation of timing of cell differentiation
embryonic digestive tract development
embryonic skeletal system development
positive regulation of epithelial cell proliferation
positive regulation of cell division
positive regulation of cardiac muscle cell proliferation
lung-associated mesenchyme development
positive regulation of canonical Wnt signaling pathway
fibroblast growth factor receptor binding
growth factor activity
heparin binding
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
osteoblast differentiation
positive regulation of mesenchymal cell proliferation
chondrocyte differentiation
protein import into nucleus
cell-cell signaling
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
male gonad development
positive regulation of gene expression
substantia nigra development
negative regulation of Wnt signaling pathway
male sex determination
lung development
embryonic limb morphogenesis
positive regulation of vascular endothelial growth factor receptor signaling pathway
positive regulation of activin receptor signaling pathway
inner ear morphogenesis
positive regulation of MAPK cascade
positive regulation of smoothened signaling pathway
regulation of timing of cell differentiation
embryonic digestive tract development
embryonic skeletal system development
positive regulation of epithelial cell proliferation
positive regulation of cell division
positive regulation of cardiac muscle cell proliferation
lung-associated mesenchyme development
positive regulation of canonical Wnt signaling pathway
fibroblast growth factor receptor binding
growth factor activity
heparin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P486914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350238 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297068 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264456 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283999 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421877 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654806 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167513 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151085 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 901030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889598 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12184 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72908 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P486914 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P486914 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P486914 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P486914 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 59349 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 70546 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72571 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 49355 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10780815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10780814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10780817 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10780816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10780818 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10780813 | - | - |
| Rat Genome 230 2.0 Array | 1392865_at | - | - |
| Rat Genome 230 2.0 Array | 1370747_at | - | - |
| Rat Genome 230 2.0 Array | 1370747_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392865_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370747_at | - | - |
| Affymetrix HT Rat Focus Array | 1392865_at | - | - |
| Affymetrix HT Rat Focus Array | 1392865_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1370747_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500004082.rn.1 | TC1500000581.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1500004083.rn.1 | TC1500000581.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370747_at | - | - |
| Rat Expression Set 230 | 1370747_PM_at | - | - |
| Rat Expression Set 230 | 1392865_at | - | - |
| Rat Expression Set 230 | 1392865_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10780813 | - | - |
| Rat Genome U34 Set | D14839_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21518483 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365421 | - | - |
| RatRef-12 Expression BeadChip | ILMN_53791 | - | - |

Fgf9
Genomic Map