Assay Details
| Gene Symbol: Ngdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107263.1 | NP_001100733.1 | 8 - 9 | 766 | 63 | |
| XM_006251966.2 | XP_006252028.1 | 8 - 9 | 959 | 63 | ||
| XM_008770676.1 | XP_008768898.1 | 8 - 9 | 1751 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ngdn
Genomic Map