Assay Details
| Gene Symbol: Atg10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109505.1 | NP_001102975.1 | 3 - 4 | 259 | 63 | |
| XM_006231756.2 | XP_006231818.1 | 4 - 5 | 526 | 63 | ||
| XM_006231757.2 | XP_006231819.1 | 3 - 4 | 1297 | 63 | ||
| XM_006231758.2 | XP_006231820.1 | 3 - 4 | 275 | 63 | ||
| XM_006231759.2 | XP_006231821.1 | 3 - 4 | 295 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Atg10
Genomic Map