Assay Details
| Gene Symbol: RGD1306227 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014037.1 | NP_001014059.1 | 5 - 6 | 1225 | 98 | |
| GenBank mRNA | BC078991.1 | - | 5 - 6 | 1225 | 7112582 | 98 |
More Information
Updated Mapping Information
Set Membership: |
|

RGD1306227
Genomic Map