Assay Details
| Gene Symbol: Ndufaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008760731.1 | XP_008758953.1 | 1 - 2 | 188 | 136 | |
| XM_342191.5 | XP_342192.1 | 1 - 2 | 186 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufaf2
Genomic Map