Assay Details
| Gene Symbol: Tmem183a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013871.1 | NP_001013893.1 | 2 - 3 | 236 | 52 | |
| XM_006249849.2 | XP_006249911.1 | 2 - 3 | 303 | 52 | ||
| XM_006249850.2 | XP_006249912.1 | 2 - 3 | 305 | 52 | ||
| GenBank mRNA | BC079375.1 | - | 2 - 3 | 236 | 7110239 | 52 |
| FQ210515.1 | - | 2 - 3 | 269 | 52 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Tmem183a
Genomic Map