Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270844.1 | NP_001257773.1 | 3 - 4 | 442 | 114 | |
| NM_001270845.1 | NP_001257774.1 | 3 - 4 | 364 | 114 | ||
| NM_182815.2 | NP_877967.2 | 3 - 4 | 364 | 114 | ||
| XM_006249713.2 | XP_006249775.1 | 3 - 4 | 457 | 114 | ||
| XM_008769436.1 | XP_008767658.1 | 2 - 3 | 379 | 114 | ||
| XM_008769437.1 | XP_008767659.1 | 3 - 4 | 380 | 114 | ||
| XM_008769438.1 | XP_008767660.1 | 3 - 4 | 363 | 114 | ||
| XM_008769439.1 | XP_008767661.1 | 3 - 4 | 458 | 114 | ||
| XM_008769440.1 | XP_008767662.1 | 3 - 4 | 550 | 114 | ||
| GenBank mRNA | AY278363.1 | - | 3 - 4 | 364 | 114 | |
| AY278364.1 | - | 3 - 4 | 364 | 114 | ||
| BC079287.1 | - | 3 - 4 | 421 | 7134311 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zp3r
Genomic Map