Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182815.2 | NP_877967.2 | 6 - 7 | 749 | 112 | |
| XM_006249713.2 | XP_006249775.1 | 6 - 7 | 842 | 112 | ||
| XM_008769436.1 | XP_008767658.1 | 5 - 6 | 764 | 112 | ||
| XM_008769437.1 | XP_008767659.1 | 6 - 7 | 765 | 112 | ||
| XM_008769438.1 | XP_008767660.1 | 6 - 7 | 748 | 112 | ||
| XM_008769440.1 | XP_008767662.1 | 6 - 7 | 935 | 112 | ||
| GenBank mRNA | AY278363.1 | - | 6 - 7 | 749 | 112 | |
More Information
Set Membership: |
|

Zp3r
Genomic Map