Assay Details
| Gene Symbol: Zp3r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270844.1 | NP_001257773.1 | 5 - 6 | 621 | 88 | |
| NM_001270845.1 | NP_001257774.1 | 5 - 6 | 543 | 88 | ||
| XM_008769439.1 | XP_008767661.1 | 5 - 6 | 637 | 88 | ||
| GenBank mRNA | AY278364.1 | - | 5 - 6 | 543 | 88 | |
| BC079287.1 | - | 5 - 6 | 600 | 7134311 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zp3r
Genomic Map