Assay Details
| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108344.2 | NP_001101814.2 | 3 - 4 | 335 | 88 | |
| GenBank mRNA | AW141189.1 | - | 3 - 4 | 335 | 88 | |
| BC167012.1 | - | 3 - 4 | 296 | 9099623 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC167012.1 transcripts.
Set Membership: |
|

Ube2t
Genomic Map