Assay Details
| Gene Symbol: Ube2t | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108344.2 | NP_001101814.2 | 2 - 3 | 266 | 103 | |
| GenBank mRNA | AW141189.1 | - | 2 - 3 | 266 | 103 | |
| BC167012.1 | - | 2 - 3 | 227 | 9099623 | 103 | |
| FQ233263.1 | - | 2 - 3 | 254 | 103 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC167012.1 transcripts.
Set Membership: |
|

Ube2t
Genomic Map