Assay Details
| Gene Symbol: Il24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133311.1 | NP_579845.1 | 2 - 3 | 401 | 127 | |
| XM_008769421.1 | XP_008767643.1 | 2 - 3 | 553 | 127 | ||
| GenBank mRNA | AF269251.1 | - | 2 - 3 | 401 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Il24
Genomic Map