Assay Details
| Gene Symbol: Fcrla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100682.1 | NP_001094152.1 | 4 - 5 | 831 | 80 | |
| GenBank mRNA | BC105902.1 | - | 4 - 5 | 802 | 80 | |
| BC158820.1 | - | 4 - 5 | 831 | 7375319 | 80 | |
| FQ225304.1 | - | 4 - 5 | 818 | 80 | ||
| FQ227783.1 | - | 4 - 5 | 820 | 80 | ||
| FQ232269.1 | - | 4 - 5 | 818 | 80 | ||
| FQ233227.1 | - | 4 - 5 | 817 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fcrla
Genomic Map