Assay Details
| Gene Symbol: Cidea | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170467.1 | NP_001163938.1 | 3 - 4 | 340 | 82 | |
| GenBank mRNA | FM041444.1 | - | 3 - 4 | 340 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cidea
Genomic Map