Assay Details
| Gene Symbol: Mc4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013099.2 | NP_037231.1 | 1 | 1810 | 156 | |
| GenBank mRNA | U67863.1 | - | 1 | 1549 | 156 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mc4r
Genomic Map