Assay Details
| Gene Symbol: Cecr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107884.1 | NP_001101354.1 | 3 - 4 | 458 | 86 | |
| XM_006237246.2 | XP_006237308.1 | 3 - 4 | 921 | 86 | ||
| XM_006237247.2 | XP_006237309.1 | 3 - 4 | 923 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cecr5
Genomic Map